APO455829345 Apollo Scientific Ltd

Ethyl 3-cyano-2-hydroxy-6-methylisonicotinate

  • MDL number MFCD00052274
  • Cas Number : 18619-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-220
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-cyano-2-hydroxy-6-methylisonicotinate

Safety Information

signalword Warning
pcodes P260;P280;P311
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-cyano-2-hydroxy-6-methylisonicotinate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart