APO455829346 Apollo Scientific Ltd

Ethyl 3-cyano-6-methyl-2-sulphanylisonicotinate

  • MDL number MFCD00052275
  • Cas Number : 56891-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-214
Synonym Ethyl 3-cyano-2-mercapto-6-methylisonicotinate, Ethyl 3-cyano-6-methyl-2-sulphanylpyridine-4-carboxylate
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23269

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23269
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart