APO455829349 Apollo Scientific Ltd

2,6-Dichloro-4-methylnicotinamide

  • MDL number MFCD00052636
  • Cas Number : 38841-54-2
  • Molecular Weight : 205.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,6-dichloro-4-methylnicotinamide
Inchi Code 1S/C7H6Cl2N2O/c1-3-2-4(8)11-6(9)5(3)7(10)12/h2H,1H3,(H2,10,12)
Inchi Key FUHRWOFCZTZGQA-UHFFFAOYSA-N

Description

Synonym 2,6-Dichloro-4-methylpyridine-3-carboxamide
Flash Point 129-132/7mm
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-4-methylnicotinamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-4-methylnicotinamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart