APO455829359 Apollo Scientific Ltd

4-tert-Butyl-2-chloroaniline

  • MDL number MFCD00052272
  • Cas Number : 42265-67-8
  • Molecular Weight : 183.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-tert-butyl-2-chloroaniline
Inchi Code 1S/C10H14ClN/c1-10(2,3)7-4-5-9(12)8(11)6-7/h4-6H,12H2,1-3H3
Inchi Key MFEZEFHCSBXPPO-UHFFFAOYSA-N

Description

Flash Point 84/0.1mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-tert-butyl-2-chloroaniline

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-tert-butyl-2-chloroaniline
pictograms GHS06
hstatements H301+H311;H315;H319;H335;H332
Availability details for
Quantity
  • Availability
  • Add to Cart