APO455829367 Apollo Scientific Ltd

Isobutanamidine hydrochloride

  • MDL number MFCD00173787
  • Cas Number : 22007-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-162
Synonym Isobutyramidine hydrochloride, 2-Methylpropanimidamide hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/isobutanamidine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/isobutanamidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart