APO455829372 Apollo Scientific Ltd

2-({2-[(2,6-Dichloropyridin-3-yl)amino]-2-oxoethyl}thio)acetic acid

  • MDL number MFCD00662316
  • Cas Number : 287197-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-{[(Carboxymethyl)sulphanyl]acetamido}-2,6-dichloropyridine, 5-({[(Carboxymethyl)thio]acetyl}amino)-2,6-dichloropyridine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23515

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23515
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart