APO455829374 Apollo Scientific Ltd

3-Hydroxy-7-methyl-1,2-benzisoxazole

  • MDL number MFCD00104669
  • Cas Number : 36238-83-2
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 155-157
Iupac Name 7-methyl-1,2-benzisoxazol-3-ol
Inchi Code 1S/C8H7NO2/c1-5-3-2-4-6-7(5)11-9-8(6)10/h2-4H,1H3,(H,9,10)
Inchi Key JUNCQBPTXAQOSA-UHFFFAOYSA-N

Description

Synonym 7-Methylbenzo[d]isoxazol-3-ol, 7-Methyl-1,2-benzisoxazol-3-ol
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 155-157
Product Link https://store.apolloscientific.co.uk/product/3-hydroxy-7-methyl-12-benzisoxazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-hydroxy-7-methyl-12-benzisoxazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart