APO455829379 Apollo Scientific Ltd

4-Bromo-1H-pyrazole-5-carboxaldehyde

  • MDL number MFCD00831607
  • Cas Number : 287917-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-180
Synonym 4-Bromo-5-formyl-1H-pyrazole
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23585

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23585
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart