APO455829385 Apollo Scientific Ltd

4-Hydroxynicotinic acid

  • MDL number MFCD00040286
  • Cas Number : 609-70-1
  • Molecular Weight : 139.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 256-259
Synonym 4-Hydroxypyridine-3-carboxylic acid, 3-Carboxy-4-hydroxypyridine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxynicotinic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxynicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart