APO455829386 Apollo Scientific Ltd

2,6-Dichloro-4-[5-(1,2-oxazol-5-yl)-1,3,4-oxadiazol-2-yl]pyridine

  • MDL number MFCD00106310
  • Cas Number : 680214-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-184
Synonym 5-[5-(2,6-Dichloropyridin-4-yl)-1,3,4-oxadiazol-2-yl]isoxazole, 2-(2,6-Dichloropyridin-4-yl)-5-(isoxazol-5-yl)-1,3,4-oxadiazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23623

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23623
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart