APO455829392 Apollo Scientific Ltd

Ethyl 2-amino-1,3-benzothiazole-6-carboxylate

  • MDL number MFCD00102724
  • Cas Number : 50850-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 241-243
Synonym 2-Amino-6-(ethoxycarbonyl)-1,3-benzothiazole
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-amino-13-benzothiazole-6-carboxylate

Safety Information

signalword Warning
pcodes P280;P271
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-amino-13-benzothiazole-6-carboxylate
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart