APO455829393 Apollo Scientific Ltd

4'-(Benzyloxy)-2'-hydroxy-3'-methylacetophenone

  • MDL number MFCD00010752
  • Cas Number : 73640-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-91
Synonym 1-[4-(Benzyloxy)-2-hydroxy-3-methylphenyl]ethan-1-one
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23655

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23655
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart