APO455829398 Apollo Scientific Ltd

N-(1-Acetyl-1H-indolin-5-yl)-4-chloro-3-nitrobenzamide

  • MDL number MFCD00277956
  • Cas Number : 680214-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Acetyl-5-[(4-chloro-3-nitrobenzoyl)amino]indoline, 5-[(4-Chloro-3-nitrobenzoyl)amino]-2,3-dihydro-1-ethanoyl-1H-indole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23694

Safety Information

signalword Warning
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23694
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart