APO455829399 Apollo Scientific Ltd

5-Hydroxy-2-nitrobenzoic acid

  • MDL number MFCD00017566
  • Cas Number : 610-37-7
  • Molecular Weight : 183.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 168-172
Iupac Name 5-hydroxy-2-nitrobenzoic acid
Inchi Code 1S/C7H5NO5/c9-4-1-2-6(8(12)13)5(3-4)7(10)11/h1-3,9H,(H,10,11)
Inchi Key BUHKQTKKZAXSMH-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 168-172
Product Link https://store.apolloscientific.co.uk/product/5-hydroxy-2-nitrobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-hydroxy-2-nitrobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart