APO455829412 Apollo Scientific Ltd

6-Amino-5-bromonicotinonitrile

  • MDL number MFCD02102412
  • Cas Number : 477871-32-2
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194-195
Synonym 6-Amino-5-bromopyridine-3-carbonitrile
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-amino-5-bromonicotinonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-amino-5-bromonicotinonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart