APO455829415 Apollo Scientific Ltd

1-(3-Chloro-4-methylphenyl)imidazolidin-2-one

  • MDL number MFCD09998747
  • Cas Number : 1031927-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(3-Chloro-4-methylphenyl)tetrahydro-2H-imidazol-2-one, 1-(3-Chloro-4-methylphenyl)-2-oxoimidazolidine, 2-Chloro-4-(2-oxoimidazolidin-1-yl)toluene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42632

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42632
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart