APO455829416 Apollo Scientific Ltd

2-(Morpholin-4-yl)-5-nitrobenzaldehyde

  • MDL number MFCD00439921
  • Cas Number : 30742-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115
Synonym 4-(2-Formyl-4-nitrophenyl)morpholine, 3-Formyl-4-(morpholin-4-yl)nitrobenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23830

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23830
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart