APO455829417 Apollo Scientific Ltd

3-[(Dimethylamino)methyl]benzeneboronic acid, pinacol ester hydrochloride

  • MDL number MFCD06657878
  • Cas Number : 1036991-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylamine hydrochloride, 2-{3-[(Dimethylamino)methyl]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane hydrochloride
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36943

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36943
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart