APO455829419 Apollo Scientific Ltd

2,6-Dimethyl-3-hydroxypyridine

  • MDL number MFCD00228692
  • Cas Number : 1122-43-6
  • Molecular Weight : 123.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-212
Synonym 2,6-Dimethylpyridin-3-ol, 3-Hydroxy-2,6-lutidine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dimethyl-3-hydroxypyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dimethyl-3-hydroxypyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart