APO455829422 Apollo Scientific Ltd

2,6-Dichloro-3-nitrobenzaldehyde

  • MDL number MFCD00090332
  • Cas Number : 5866-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-74
Synonym 2,4-Dichloro-3-formylnitrobenzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-3-nitrobenzaldehyde

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-3-nitrobenzaldehyde
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart