APO455829423 Apollo Scientific Ltd

2,6-Dichloro-3-nitrobenzyl alcohol

  • MDL number MFCD01566901
  • Cas Number : 160647-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-108
Synonym (2,6-Dichloro-3-nitrophenyl)methanol, 2,4-Dichloro-3-(hydroxymethyl)nitrobenzene
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23890

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23890
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart