APO455829426 Apollo Scientific Ltd

1-(Chloromethyl)cyclohexanecarbonitrile

  • MDL number MFCD01570824
  • Cas Number : 112905-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 83-86/0.2mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23898

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23898
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart