APO455829431 Apollo Scientific Ltd

Methyl 3-methyl-2-nitrobenzoate

  • MDL number MFCD01003947
  • Cas Number : 5471-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-76
Synonym 2-(Methoxycarbonyl)-6-methylnitrobenzene, Methyl 2-nitro-m-toluate
Purity 98
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23941

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23941
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart