APO455829433 Apollo Scientific Ltd

2-Methoxypyrimidine-5-carboxaldehyde

  • MDL number MFCD03446732
  • Cas Number : 90905-32-1
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Formyl-2-methoxypyrimidine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methoxypyrimidine-5-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxypyrimidine-5-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart