APO455829439 Apollo Scientific Ltd

Ethyl 3-oxocyclohexane-1-carboxylate

  • MDL number MFCD00205583
  • Cas Number : 33668-25-6
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-oxocyclohexanecarboxylate
density 1.068
Inchi Code 1S/C9H14O3/c1-2-12-9(11)7-4-3-5-8(10)6-7/h7H,2-6H2,1H3
Inchi Key YLRVJPQVDQQBOX-UHFFFAOYSA-N

Description

Synonym 1-(Ethoxycarbonyl)-3-oxocyclohexane, 3-(Ethoxycarbonyl)cyclohexan-1-one
Purity 97%
Density 1.068
Flash Point 135-137/15mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-oxocyclohexane-1-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-oxocyclohexane-1-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart