APO455829452 Apollo Scientific Ltd

4-(N,N-Diethylamino)phenacyl bromide

  • MDL number MFCD00151798
  • Cas Number : 207986-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52
Synonym 2-Bromo-1-[4-(diethylamino)phenyl]ethan-1-one
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=24114

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=24114
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart