APO455829455 Apollo Scientific Ltd

3-Chloroisonicotinonitrile

  • MDL number MFCD05663706
  • Cas Number : 68325-15-5
  • Molecular Weight : 138.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-74
Synonym 3-Chloro-4-cyanopyridine, 3-Chloropyridine-4-carbonitrile
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloroisonicotinonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloroisonicotinonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart