APO455829467 Apollo Scientific Ltd

Methyl 2,4-dioxo-4-(2-nitrophenyl)butanoate

  • MDL number MFCD00832016
  • Cas Number : 57446-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 2,4-dioxo-4-(2-nitrophenyl)butyrate, 2-(4-Methoxy-3,4-dioxobutanoyl)nitrobenzene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=24318

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=24318
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart