APO455829474 Apollo Scientific Ltd

Methyl 3,6-dichloropyridine-2-carboxylate

  • MDL number MFCD00186642
  • Cas Number : 1532-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 3,6-dichloropicolinate, 3,6-Dichloro-2-(methoxycarbonyl)pyridine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=24365

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=24365
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart