APO455829477 Apollo Scientific Ltd

1-(4-Benzylpiperazin-1-yl)-2-(5-nitro-1H-indol-3-yl)ethane-1,2-dione

  • MDL number MFCD01935748
  • Cas Number : 334973-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-[(4-Benzylpiperazin-1-yl)(oxo)acetyl]-5-nitro-1H-indole, 1-Benzyl-4-[(5-nitro-1H-indol-3-yl)(oxo)acetyl]piperazine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=24386

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=24386
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart