APO455829482 Apollo Scientific Ltd

Allylamine, N-CBZ protected

  • MDL number MFCD01570995
  • Cas Number : 5041-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Benzyl allylcarbamate, Benzyl (prop-2-en-1-yl)carbamate
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 120-125/1mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=24471

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=24471
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart