APO455829485 Apollo Scientific Ltd

Ethyl 4-methyl-1,3-oxazole-5-carboxylate

  • MDL number MFCD00062573
  • Cas Number : 20485-39-6
  • Molecular Weight : 155.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 36-38
Iupac Name ethyl 4-methyl-1,3-oxazole-5-carboxylate
Inchi Code 1S/C7H9NO3/c1-3-10-7(9)6-5(2)8-4-11-6/h4H,3H2,1-2H3
Inchi Key XNMORZSEENWFLI-UHFFFAOYSA-N

Description

Synonym 5-(Ethoxycarbonyl)-4-methyl-1,3-oxazole
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 36-38
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-methyl-13-oxazole-5-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-methyl-13-oxazole-5-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart