APO455829496 Apollo Scientific Ltd

1-(Cyclopropylmethyl)thiourea

  • MDL number MFCD00276905
  • Cas Number : 618913-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-113
Synonym [(Carbamothioylamino)methyl]cyclopropane, (Thioureidomethyl)cyclopropane
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=24545

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=24545
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart