APO455829500 Apollo Scientific Ltd

4-Hydroxy-6-methylquinazoline

  • MDL number MFCD00174855
  • Cas Number : 19181-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238-240
Synonym 6-Methylquinazolin-4-ol, 6-Methylquinazolin-4(3H)-one
Purity 97
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-6-methylquinazoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-6-methylquinazoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart