APO455829513 Apollo Scientific Ltd

1,4-Bis(4-phenoxybenzoyl)benzene

  • MDL number MFCD00319174
  • Cas Number : 54299-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-218
Synonym Phenylene-1,4-bis[(4-phenoxyphenyl)methanone]
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37393

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37393
pictograms GHS07
hstatements H315;H317;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart