APO455829514 Apollo Scientific Ltd

N-(3,4-Dichlorophenyl)thiourea

  • MDL number MFCD00004935
  • Cas Number : 19250-09-0
  • Molecular Weight : 221.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197-200(dec.)
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-34-dichlorophenylthiourea

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-34-dichlorophenylthiourea
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart