APO455829529 Apollo Scientific Ltd

Methyl 4-amino-3-methoxybenzoate

  • MDL number MFCD00017203
  • Cas Number : 41608-64-4
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 128-131
Iupac Name methyl 4-amino-3-methoxybenzoate
Inchi Code 1S/C9H11NO3/c1-12-8-5-6(9(11)13-2)3-4-7(8)10/h3-5H,10H2,1-2H3
Inchi Key DJLFOMMCQBAMAA-UHFFFAOYSA-N

Description

Synonym 2-Methoxy-4-(methoxycarbonyl)aniline, 2-Amino-5-(methoxycarbonyl)anisole
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 128-131
Product Link https://store.apolloscientific.co.uk/product/methyl-4-amino-3-methoxybenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-amino-3-methoxybenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart