APO455829549 Apollo Scientific Ltd

2,3-Dimethyl-6-nitroaniline

  • MDL number MFCD00010148
  • Cas Number : 59146-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-122
Synonym 6-Nitro-2,3-xylidine, 2-Amino-3,4-dimethylnitrobenzene, 3-Amino-4-nitro-o-xylene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=24817

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=24817
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart