APO455829550 Apollo Scientific Ltd

6-Phenylpyridine-2-carboxylic acid

  • MDL number MFCD04114206
  • Cas Number : 39774-28-2
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 95-100
Iupac Name 6-phenyl-2-pyridinecarboxylic acid
Inchi Code 1S/C12H9NO2/c14-12(15)11-8-4-7-10(13-11)9-5-2-1-3-6-9/h1-8H,(H,14,15)
Inchi Key CXOPUGLYEYDAMC-UHFFFAOYSA-N

Description

Synonym 6-Phenylpicolinic acid, 2-Carboxy-6-phenylpyridine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 95-100
Product Link https://store.apolloscientific.co.uk/product/6-phenylpyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-phenylpyridine-2-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart