APO455829554 Apollo Scientific Ltd

4-Chloro-2-cyclohexylphenol

  • MDL number MFCD00019337
  • Cas Number : 13081-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-60
Synonym 1-Chloro-3-cyclohexyl-4-hydroxybenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 170-180/18mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=24835

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=24835
pictograms GHS05;GHS06;GHS07
hstatements H301;H318;H335;H413
Availability details for
Quantity
  • Availability
  • Add to Cart