APO455829559 Apollo Scientific Ltd

4'-Isopropylacetophenone

  • MDL number MFCD00048297
  • Cas Number : 645-13-6
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[4-(Prop-2-yl)phenyl]ethan-1-one, 4'-(Prop-2-yl)acetophenone
Density 0.97
Flash Point >110
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 238
Product Link https://store.apolloscientific.co.uk/product/4-isopropylacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-isopropylacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart