APO455829568 Apollo Scientific Ltd

2-Chloro-6-iodotoluene

  • MDL number MFCD00019013
  • Cas Number : 42048-11-3
  • Molecular Weight : 252.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 11
Iupac Name 1-chloro-3-iodo-2-methylbenzene
density 1.82
Inchi Code 1S/C7H6ClI/c1-5-6(8)3-2-4-7(5)9/h2-4H,1H3
Inchi Key OEHHXVIJMCMYGM-UHFFFAOYSA-N

Description

Synonym 1-Chloro-3-iodo-2-methylbenzene
Density 1.82
Flash Point 117-119
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 11
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-iodotoluene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-iodotoluene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart