APO455829569 Apollo Scientific Ltd

Methyl 2-(chlorosulphonyl)benzoate

  • MDL number MFCD00009797
  • Cas Number : 26638-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-64
Synonym 2-(Methoxycarbonyl)benzenesulphonyl chloride
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-2-chlorosulphonylbenzoate

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-chlorosulphonylbenzoate
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart