APO455829576 Apollo Scientific Ltd

Ethyl 4,6-dihydroxynicotinate

  • MDL number MFCD00834976
  • Cas Number : 6975-44-6
  • Molecular Weight : 183.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-217
Synonym Ethyl 4,6-dihydroxypyridine-3-carboxylate, 2,4-Dihydroxy-5-(ethoxycarbonyl)pyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-46-dihydroxynicotinate

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-46-dihydroxynicotinate
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart