APO455829595 Apollo Scientific Ltd

Adamantane-1-carboxylic acid

  • MDL number MFCD00074720
  • Cas Number : 828-51-3
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 172-176
Iupac Name 1-adamantanecarboxylic acid
Inchi Code 1S/C11H16O2/c12-10(13)11-4-7-1-8(5-11)3-9(2-7)6-11/h7-9H,1-6H2,(H,12,13)
Inchi Key JIMXXGFJRDUSRO-UHFFFAOYSA-N

Description

Synonym 1-Carboxyadamantane, Tricyclo[3.3.1.1~3,7~]decane-1-carboxylic acid
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 172-176
Product Link https://store.apolloscientific.co.uk/product/adamantane-1-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/adamantane-1-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart