APO455829615 Apollo Scientific Ltd

4-Acetyl-6-(tert-butyl)-1,1,-dimethylindane

  • MDL number MFCD00046324
  • Cas Number : 13171-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-79
Synonym Celestolide, 1-[6-(tert-Butyl)-2,3-dihydro-1,1-dimethyl-1H-inden-4-yl]ethan-1-one
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-acetyl-6-tert-butyl-11-dimethylindane

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-acetyl-6-tert-butyl-11-dimethylindane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart