APO455829618 Apollo Scientific Ltd

2-(Bromoacetyl)-1,3-thiazole

  • MDL number MFCD06411540
  • Cas Number : 3292-77-1
  • Molecular Weight : 206.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 54-55
Iupac Name 2-bromo-1-(1,3-thiazol-2-yl)ethanone
Inchi Code 1S/C5H4BrNOS/c6-3-4(8)5-7-1-2-9-5/h1-2H,3H2
Inchi Key AQRFTRDAOYSMEA-UHFFFAOYSA-N

Description

Synonym 2-Bromo-1-(1,3-thiazol-2-yl)ethan-1-one, 2-(1-Bromoethanoyl)-1,3-thiazole
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 54-55
Product Link https://store.apolloscientific.co.uk/product/2-bromoacetyl-13-thiazole

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS https://store.apolloscientific.co.uk/product/2-bromoacetyl-13-thiazole
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart