APO455829633 Apollo Scientific Ltd

2,4-Dimethylphenyl isothiocyanate

  • MDL number MFCD00022054
  • Cas Number : 39842-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 25-29
Synonym 4-Isothiocyanato-m-xylene, 2,4-Dimethyl-1-isothiocyanatobenzene
Storage temp. Ambient temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 122-130/8mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25042

Safety Information

signalword Danger
pcodes P260;P280;P301+P310
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25042
pictograms GHS05;GHS06;GHS08
hstatements H301;H314;H317;H332;H334
Availability details for
Quantity
  • Availability
  • Add to Cart