APO455829637 Apollo Scientific Ltd

6,7-Dimethoxy-1-(3,4,5-triethoxyphenyl)isoquinoline

  • MDL number MFCD00118512
  • Cas Number : 549-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-103
Synonym 5-(6,7-Dimethoxyisoquinolin-1-yl)-1,2,3-triethoxybenzene, 6,7-Dimethoxy-1-(3,4,5-triethoxyphenyl)-2-azanaphthalene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25053

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25053
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart