APO455829646 Apollo Scientific Ltd

2-[2-(Dimethylamino)ethyl]pyridine

  • MDL number MFCD00023546
  • Cas Number : 6304-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N-Dimethyl-2-(pyridin-2-yl)ethylamine, N-[2-(Pyridin-2-yl)ethyl]dimethylamine
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25073

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25073
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart